Merck
  • Metformin induces up-regulation of blood-brain barrier functions by activating AMP-activated protein kinase in rat brain microvascular endothelial cells.
Metformin induces up-regulation of blood-brain barrier functions by activating AMP-activated protein kinase in rat brain microvascular endothelial cells.
Biochemical and biophysical research communications (2013-03-26)
Fuyuko Takata, Shinya Dohgu, Junichi Matsumoto, Takashi Machida, Shuji Kaneshima, Mai Matsuo, Shinya Sakaguchi, Yuki Takeshige, Atsushi Yamauchi, Yasufumi Kataoka
ZUSAMMENFASSUNG
Blood-brain barrier (BBB) disruption occurs frequently in CNS diseases and injuries. Few drugs have been developed as therapeutic candidates for facilitating BBB functions. Here, we examined whether metformin up-regulates BBB functions using rat brain microvascular endothelial cells (RBECs). Metformin, concentration- and time-dependently increased transendothelial electrical resistance of RBEC monolayers, and decreased RBEC permeability to sodium fluorescein and Evans blue albumin. These effects of metformin were blocked by compound C, an inhibitor of AMP-activated protein kinase (AMPK). AMPK stimulation with an AMPK activator, AICAR, enhanced BBB functions. These findings indicate that metformin induces up-regulation of BBB functions via AMPK activation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Fluoreszein Natriumsalz, BioReagent, suitable for fluorescence
Sigma-Aldrich
Fluorescein, free acid
Sigma-Aldrich
Fluoreszein Natriumsalz, used as fluorescent tracer
Sigma-Aldrich
Fluorescein (freie Säure), Dye content 95 %
Sigma-Aldrich
Fluoreszein Natriumsalz, p.a.
Sigma-Aldrich
Myokinase aus Kaninchenmuskel, ammonium sulfate suspension, 1,500-3,000 units/mg protein (biuret)