Merck
Metformin retards aging in C. elegans by altering microbial folate and methionine metabolism.
Cell (2013-04-02)
Filipe Cabreiro, Catherine Au, Kit-Yi Leung, Nuria Vergara-Irigaray, Helena M Cochemé, Tahereh Noori, David Weinkove, Eugene Schuster, Nicholas D E Greene, David Gems
ZUSAMMENFASSUNG
The biguanide drug metformin is widely prescribed to treat type 2 diabetes and metabolic syndrome, but its mode of action remains uncertain. Metformin also increases lifespan in Caenorhabditis elegans cocultured with Escherichia coli. This bacterium exerts complex nutritional and pathogenic effects on its nematode predator/host that impact health and aging. We report that metformin increases lifespan by altering microbial folate and methionine metabolism. Alterations in metformin-induced longevity by mutation of worm methionine synthase (metr-1) and S-adenosylmethionine synthase (sams-1) imply metformin-induced methionine restriction in the host, consistent with action of this drug as a dietary restriction mimetic. Metformin increases or decreases worm lifespan, depending on E. coli strain metformin sensitivity and glucose concentration. In mammals, the intestinal microbiome influences host metabolism, including development of metabolic disease. Thus, metformin-induced alteration of microbial metabolism could contribute to therapeutic efficacy-and also to its side effects, which include folate deficiency and gastrointestinal upset.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
L-Methionin, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
SAFC
L-Methionin
Sigma-Aldrich
L-Methionin, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Methionin, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
DL-Methionin, ≥99%
Supelco
L-Methionin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
DL-Methionin, 99%, FCC, FG
Sigma-Aldrich
Myokinase aus Kaninchenmuskel, ammonium sulfate suspension, 1,500-3,000 units/mg protein (biuret)
Methionin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
DL-Methionin, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
DL-Methionin, ≥99.0% (NT)
Supelco
L-Methionin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland