Merck
Effect of Chinese herbs on immunoglobulin A nephropathy: a randomized controlled trial.
Journal of traditional Chinese medicine = Chung i tsa chih ying wen pan (2013-04-20)
Minggang Wei, Peihua Xiong, Ling Zhang
ZUSAMMENFASSUNG
The accumulation of extracellular matrix (ECM) is one of the main causes of renal fibrosis. Emerging evidence suggests that the metabolic enzyme of ECM is associated with renal fibrosis. In this study, we applied randomly controlled trial to check the curative effect of Chinese herbs on patients with immunoglobulin A nephropathy (IgAN). Twenty-six patients were randomly divided into group A (control group) treated with Western Medicine and group B (treatment group) treated with combination of Traditional Chinese Medicine (TCM) and Western Medicine. Blood and urine tests were done before treatment and after 8-week treatment. The levels of the main composition of extracellular matrix (MC-ECM), the metabolic enzyme of ECM (ME-ECM) and some cytokines in group B decreased more obviously than those in group A after 8-week treatment. So did the level of 24-hour urine protein. However, Metal matrix protease (MMP)-2 and vascular endothelial growth factor in group B increased more obviously than those in group A after 8-week treatment. No effects on the renal function were found in both groups. Our study provided important information on using the combination of TCM with Western Medicine to inhibit the progression of renal fibrosis in patients with IgAN.