Merck
  • Amino acid sequence requirement for efficient incorporation of glycosylphosphatidylinositol-associated proteins into the cell wall of Saccharomyces cerevisiae.
Amino acid sequence requirement for efficient incorporation of glycosylphosphatidylinositol-associated proteins into the cell wall of Saccharomyces cerevisiae.
The Journal of biological chemistry (1998-10-03)
K Hamada, H Terashima, M Arisawa, K Kitada
ZUSAMMENFASSUNG
During cell wall biogenesis in Saccharomyces cerevisiae, some glycosylphosphatidylinositol (GPI)-attached proteins are detached from GPI moieties and bound to beta-1,6-glucan of the cell wall. The amino acid sequence requirement for the incorporation of GPI-attached proteins into the cell wall was studied by using reporter fusion proteins. Only the short omega-minus region composed of five amino acids, which is located upstream of the omega site for GPI attachment, determined the cellular localization of the GPI-associated proteins. Within the omega-minus region, amino acid residues at the omega-4 or -5 and omega-2 sites were important for the cell wall incorporation. Yap3p, a well characterized GPI-anchored plasma membrane aspartic protease, was localized in the cell wall when the omega-minus region was mutated to sequences containing Val or Ile at the omega-4 or -5 site and Val or Tyr at the omega-2 site.