Merck
Removal of N-terminal blocking groups from proteins.
Current protocols in protein science (2011-03-15)
Joseph W Leone, Brian Hampton, Elizabeth Fowler, Mary Moyer, Radha G Krishna, Christopher C Q Chin
ZUSAMMENFASSUNG
Two enzymatic methods commonly used in N-terminal sequence analysis of blocked proteins are presented: one uses pyroglutamate aminopeptidase for N(α)-pyrrolidone carboxyl-proteins in solution or blotted onto a membrane, and the other uses acylaminoacyl-peptide hydrolase for N(α)-acyl-proteins blocked with other acyl groups. A Support Protocol describes a colorimetric assay for pyroglutamate aminopeptidase activity. Sequencing with acylaminoacyl-peptide hydrolase must include fragmentation of the protein before unblocking, so procedures are provided for chemically blocking newly generated peptides with either succinic anhydride or phenylisothiocyanate/performic acid. The hydrolase is then applied to the total mixture of peptides, only one of which, the acylated N-terminal peptide, should be a substrate for hydrolase. After incubation, the mixture of peptides is subjected to sequence analysis.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Bernsteinsäureanhydrid, ≥99% (GC)
Sigma-Aldrich
Phenylisothiocyanat, reagent grade, 98%
Sigma-Aldrich
Phenylisothiocyanat, for HPLC derivatization, the detection of alcohols and amines, ≥99.0%
Sigma-Aldrich
Phenylisothiocyanat, 99%, for protein sequencing
Sigma-Aldrich
Pyroglutamate Aminopeptidase from Pyrococcus furiosus, recombinant, expressed in E. coli, ~90% (SDS-PAGE), ≥5.0 units/mg protein
Sigma-Aldrich
Phenylisothiocyanat, Sigma Grade, 8.36 M, suitable for solid phase protein sequencing analysis, ≥99% (GC), liquid