Merck
  • Photosynthetic responses of a C(3) and three C(4) species of the genus Panicum (s.l.) with different metabolic subtypes to drought stress.
Photosynthetic responses of a C(3) and three C(4) species of the genus Panicum (s.l.) with different metabolic subtypes to drought stress.
Photosynthesis research (2012-07-17)
Sabrina U Alfonso, Wolfgang Brüggemann
ZUSAMMENFASSUNG
Young plants of Panicum bisulcatum (C(3)), Zuloagaea bulbosa [NADP-malic enzyme (ME)-C(4)], P. miliaceum (NAD-ME-C(4)) and Urochloa maxima [phosphoenolpyruvate carboxykinase (PCK)-C(4)] were subjected to drought stress (DS) in soil for 6 days. The C(3) species showed severe wilting symptoms at higher soil water potential (-1.1 MPa) and relative leaf water content (77 %) than in the case of the C(4) species (-1.5 to -1.7 MPa; 58-64 %). DS decreased photosynthesis, both under atmospheric and under saturating CO(2). Stomatal limitation of net photosynthesis (P(N)) in the C(3), but not in the C(4) species was indicated by P(N)/C(o) curves. Chlorophyll fluorescence of photosystem II, resulting from different cell types in the four species, indicated NADPH accumulation and non-stomatal limitation of photosynthesis in all four species, even under high CO(2). In the NAD-ME-C(4) and the PCK-C(4) species, DS plants showed increased violaxanthin de-epoxidase rates. Biochemical analyses of carboxylating enzymes and in vitro enzyme activities of the C(4) enzymes identified the most likely non-stomatal limiting steps of photosynthesis. In P. bisulcatum, declining RubisCO content and activity would explain the findings. In Z. bulbosa, all photosynthesis enzymes declined significantly; photosynthesis is probably limited by the turnover rate of the PEPC reaction. In P. miliaceum, all enzyme levels remained fairly constant under DS, but photosynthesis can be limited by feedback inhibition of the Calvin cycle, resulting in asp inhibition of PEPC. In U. maxima, declines of in vivo PEPC activity and feedback inhibition of the Calvin cycle are the main candidates for non-stomatal limitation of photosynthesis under DS.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Phosphoenolpyruvate carboxylase microbial, ≥5.0 unit/mg solid