Merck
  • Three-dimensional cultures modeling premalignant progression of human breast epithelial cells: role of cysteine cathepsins.
Three-dimensional cultures modeling premalignant progression of human breast epithelial cells: role of cysteine cathepsins.
Biological chemistry (2013-05-15)
Stefanie R Mullins, Mansoureth Sameni, Galia Blum, Matthew Bogyo, Bonnie F Sloane, Kamiar Moin
ZUSAMMENFASSUNG
The expression of the cysteine protease cathepsin B is increased in early stages of human breast cancer.To assess the potential role of cathepsin B in premalignant progression of breast epithelial cells, we employed a 3D reconstituted basement membrane overlay culture model of MCF10A human breast epithelial cells and isogenic variants that replicate the in vivo phenotypes of hyper plasia(MCF10AneoT) and atypical hyperplasia (MCF10AT1). MCF10A cells developed into polarized acinar structures with central lumens. In contrast, MCF10AneoT and MCF10AT1 cells form larger structures in which the lumens are filled with cells. CA074Me, a cell-permeable inhibitor selective for the cysteine cathepsins B and L,reduced proliferation and increased apoptosis of MCF10A, MCF10AneoT and MCF10AT1 cells in 3D culture. We detected active cysteine cathepsins in the isogenic MCF10 variants in 3D culture with GB111, a cell-permeable activity based probe, and established differential inhibition of cathepsin B in our 3D cultures. We conclude that cathepsin B promotes proliferation and premalignant progression of breast epithelial cells. These findings are consistent with studies by others showing that deletion of cathepsin B in the transgenic MMTV-PyMT mice, a murine model that is predisposed to development of mammary cancer, reduces malignant progression.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
L-Leucin, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Leucin, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
Cathepsin B from human liver, buffered aqueous solution, ≥200 units/mg protein (One unit is defined as the amount of enzyme that hydrolyzes 1 μmol of Z-Arg-Arg-beta-naphthylamide per minute at 40 °C, using 100 mM Na/K phosphate, pH 6.0, with 1.33 mM EDTA and 2 mM DTT as the activation buffer.)
Sigma-Aldrich
L-Leucin, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Cathepsin B aus Rindermilz, lyophilized powder, ≥10 units/mg protein
Supelco
L-Leucin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Cathepsin B from human placenta, lyophilized powder, ≥5 units/mg protein
Sigma-Aldrich
L-Leucin, 99%, FG
Sigma-Aldrich
Cathepsin L from human liver, ≥0.5 units/mg protein, solution
Supelco
L-Leucin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland