Merck
  • Mutant p53 prolongs NF-κB activation and promotes chronic inflammation and inflammation-associated colorectal cancer.
Mutant p53 prolongs NF-κB activation and promotes chronic inflammation and inflammation-associated colorectal cancer.
Cancer cell (2013-05-18)
Tomer Cooks, Ioannis S Pateras, Ohad Tarcic, Hilla Solomon, Aaron J Schetter, Sylvia Wilder, Guillermina Lozano, Eli Pikarsky, Tim Forshew, Nitzan Rosenfeld, Nitzan Rozenfeld, Noam Harpaz, Steven Itzkowitz, Curtis C Harris, Varda Rotter, Vassilis G Gorgoulis, Moshe Oren
ZUSAMMENFASSUNG
The tumor suppressor p53 is frequently mutated in human cancer. Common mutant p53 (mutp53) isoforms can actively promote cancer through gain-of-function (GOF) mechanisms. We report that mutp53 prolongs TNF-α-induced NF-κB activation in cultured cells and intestinal organoid cultures. Remarkably, when exposed to dextran sulfate sodium, mice harboring a germline p53 mutation develop severe chronic inflammation and persistent tissue damage, and are highly prone to inflammation-associated colon cancer. This mutp53 GOF is manifested by rapid onset of flat dysplastic lesions that progress to invasive carcinoma with mutp53 accumulation and augmented NF-κB activation, faithfully recapitulating features frequently observed in human colitis-associated colorectal cancer (CAC). These findings might explain the early appearance of p53 mutations in human CAC.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Azoxymethan, 13.4 M, ≥98%
Sigma-Aldrich
Nitric Oxide Synthase, Inducible from mouse, recombinant, expressed in E. coli, buffered aqueous solution