Merck
  • Effects of corn replacement by sorghum in broiler diets on performance and intestinal mucosa integrity.
Effects of corn replacement by sorghum in broiler diets on performance and intestinal mucosa integrity.
Poultry science (2013-05-21)
K A A Torres, J M Pizauro, C P Soares, T G A Silva, W C L Nogueira, D M B Campos, R L Furlan, M Macari
ZUSAMMENFASSUNG
The effect of replacing corn with low-tannin sorghum on broiler performance, carcass yield, integrity of mucosa of small intestine segments, and activity of membrane enzymes of the jejunum is investigated. A total of 594 male Cobb-500 broiler chicks were randomly assigned to 3 dietary treatments: 100% corn (control), 50% corn replacement with low-tannin sorghum (low sorghum), and 100% corn replacement with low-tannin sorghum (high sorghum). Body weight gain, feed consumption, feed conversion, and carcass yield were determined at 7, 21, and 42 d, and segments of the small intestine were collected. Feed conversion and weight gain were impaired at d 42 in broilers fed the high-sorghum diet, but no differences were observed for carcass yield among the treatments (P > 0.05). Crypt cell mitotic index of the jejunum and ileum at d 21 and 42 was lower in broilers fed the control diet than in those fed low- and high-sorghum diets (P < 0.05). Aminopeptidase activity was higher in broilers fed the control diet than in those fed low- and high-sorghum diets irrespective of age (P < 0.05). Conversely, intestinal alkaline phosphatase activity in the small intestine did not differ among the dietary treatments (P > 0.05). Our results indicate that 50% corn replacement with low-tannin sorghum is suitable for broiler diets, whereas 100% corn replacement with low-tannin sorghum had negative effects on the intestinal mucosa and performance of broilers at 42 d.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Phosphatase, alkalisch aus Rinderdarmschleimhaut, buffered aqueous solution, ≥2,000 DEA units/mg protein
Sigma-Aldrich
Phosphatase, alkalisch aus Rinderdarmschleimhaut, lyophilized powder, ≥10 DEA units/mg solid
Sigma-Aldrich
Phosphatase, alkalisch aus Rinderdarmschleimhaut, BioUltra, ≥5,700 DEA units/mg protein
Sigma-Aldrich
Phosphatase, alkalisch aus E. coli, lyophilized powder, 30-60 units/mg protein (in glycine buffer)
Sigma-Aldrich
Phosphatase, alkalisch aus Rinderdarmschleimhaut, ≥5,500 DEA units/mg protein
Sigma-Aldrich
Phosphatase, alkalisch aus E. coli, buffered aqueous glycerol solution, 20-50 units/mg protein (in glycine buffer)
Sigma-Aldrich
Phosphatase, alkalisch aus Rinderdarmschleimhaut, buffered aqueous glycerol solution, ≥4,000 DEA units/mg protein
Sigma-Aldrich
Phosphatase, alkalisch aus E. coli, ammonium sulfate suspension, 30-90 units/mg protein (modified Warburg-Christian, in glycine buffer)
Sigma-Aldrich
Phosphatase, alkalisch aus Schweineniere, lyophilized powder, ≥100 DEA units/mg protein
Sigma-Aldrich
Phosphatase, alkalisch aus Kälberdarmschleimhaut, suitable for enzyme immunoassay, solution (clear, colorless), ~2500 U/mg protein (~10 mg/ml)
Sigma-Aldrich
Phosphatase, alkalische Garnelen, ≥900 DEA units/mL, buffered aqueous glycerol solution, recombinant, expressed in proprietary host
Sigma-Aldrich
Phosphatase, alkalisch Rind, recombinant, expressed in Pichia pastoris, ≥4000 units/mg protein