Merck
  • Highly-sensitive cholesterol biosensor based on platinum-gold hybrid functionalized ZnO nanorods.
Highly-sensitive cholesterol biosensor based on platinum-gold hybrid functionalized ZnO nanorods.
Talanta (2012-05-23)
Chengyan Wang, Xingrong Tan, Shihong Chen, Ruo Yuan, Fangxin Hu, Dehua Yuan, Yun Xiang
ZUSAMMENFASSUNG
A novel scheme for the fabrication of gold/platinum hybrid functionalized ZnO nanorods (Pt-Au@ZnONRs) and multiwalled carbon nanotubes (MWCNTs) modified electrode is presented and its application for cholesterol biosensor is investigated. Firstly, Pt-Au@ZnONRs was prepared by the method of chemical synthesis. Then, the Pt-Au@ZnONRs suspension was dropped on the MWCNTs modified glass carbon electrode, and followed with cholesterol oxidase (ChOx) immobilization by the adsorbing interaction between the nano-material and ChOx as well as the electrostatic interaction between ZnONRs and ChOx molecules. The combination of MWCNTs and Pt-Au@ZnONRs provided a favorable environment for ChOx and resulted in the enhanced analytical response of the biosensor. The resulted biosensor exhibited a linear response to cholesterol in the wide range of 0.1-759.3 μM with a low detection limit of 0.03 μM and a high sensitivity of 26.8 μA mM(-1). The calculated apparent Michaelis constant K(M)(app) was 1.84 mM, indicating a high affinity between ChOx and cholesterol.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Cholesterol Oxidase from Brevibacterium, lyophilized powder, ≥50 units/mg protein, recombinant, expressed in E. coli
Sigma-Aldrich
Cholesterin-Oxidase aus Streptomyces sp., lyophilized powder, ≥20 units/mg protein
Sigma-Aldrich
Cholesterol Oxidase microbial, recombinant, lyophilized powder, ≥10 units/mg protein
Sigma-Aldrich
Cholesterol Oxidase from microorganisms, aqueous solution, ≥30 units/mg protein (biuret)