Merck
  • Sesamin is a potent and specific inhibitor of delta 5 desaturase in polyunsaturated fatty acid biosynthesis.
Sesamin is a potent and specific inhibitor of delta 5 desaturase in polyunsaturated fatty acid biosynthesis.
Lipids (1991-07-01)
S Shimizu, K Akimoto, Y Shinmen, H Kawashima, M Sugano, H Yamada
ZUSAMMENFASSUNG
Incubation with sesame oil increases the mycelial dihomo-gamma-linolenic acid content of an arachidonic acid-producing fungus, Mortierella alpina, but decreases its arachidonic acid content [Shimizu, S., K. Akimoto, H. Kawashima, Y. Shinmen and H. Yamada (1989) J. Am. Oil Chem. Soc. 66, 237-241]. The factor causing these effects was isolated and identified to be (+)-sesamin. The results obtained in experiments with both a cell-free extract of the fungus and with rat liver microsomes demonstrated that (+)-sesamin specifically inhibits delta 5 desaturase at low concentrations, but does not inhibit delta 6, delta 9 and delta 12 desaturases. Kinetic analysis showed that (+)-sesamin is a noncompetitive inhibitor (Ki for rat liver delta 5 desaturase, 155 microM). (+)-Sesamolin, (+)-sesaminol and (+)-episesamin also inhibited only delta 5 desaturases of the fungus and liver. These results demonstrate that (+)-sesamin and related lignan compounds present in sesame seeds or its oil are specific inhibitors of delta 5 desaturase in polyunsaturated fatty acid biosynthesis in both microorganisms and animals.