Merck
  • Identification of a novel protein, PDIP38, that interacts with the p50 subunit of DNA polymerase delta and proliferating cell nuclear antigen.
Identification of a novel protein, PDIP38, that interacts with the p50 subunit of DNA polymerase delta and proliferating cell nuclear antigen.
The Journal of biological chemistry (2003-01-11)
Li Liu, Esther M Rodriguez-Belmonte, Nayef Mazloum, Bin Xie, Marietta Y W T Lee
ZUSAMMENFASSUNG
The yeast two-hybrid screening method was used to identify novel proteins that associate with human DNA polymerase delta (pol delta). Two baits were used in this study. These were the large (p125) and small (p50) subunits of the core pol delta heterodimer. p50 was the only positive isolated with p125 as the bait. Two novel protein partners, named PDIP38 and PDIP46, were identified from the p50 screen. In this study, the interaction of PDIP38 with pol delta was further characterized. PDIP38 encodes a protein of 368 amino acids whose C terminus is conserved with the bacterial APAG protein and with the F box A protein. It was found that PDIP38 also interacts with proliferating cell nuclear antigen (PCNA). The ability of PDIP38 to interact with both the p50 subunit of pol delta and with PCNA was confirmed by pull-down assays using glutathione S-transferase (GST)-PDIP38 fusion proteins. The PCNA-PDIP38 interaction was also demonstrated by PCNA overlay experiments. The association of PDIP38 with pol delta was shown to occur in calf thymus tissue and mammalian cell extracts by GST-PDIP38 pull-down and coimmunoprecipitation experiments. PDIP38 was associated with pol delta isolated by immunoaffinity chromatography. The association of PDIP38 with pol delta could also be demonstrated by native gel electrophoresis.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Taq-DNA-Polymerase aus Thermus aquaticus, with 10× PCR reaction buffer containing MgCl2
Sigma-Aldrich
Taq-DNA-Polymerase aus Thermus aquaticus, with 10× PCR reaction buffer without MgCl2
Sigma-Aldrich
DNA-Polymerase I aus E. coli lysogen für NM 964, buffered aqueous glycerol solution