Merck
cDNA sequence of the long mRNA for human glutamine synthase.
Biochimica et biophysica acta (1991-10-08)
M J Van den Hoff, W J Geerts, A T Das, A F Moorman, W H Lamers
ZUSAMMENFASSUNG
Screening a human liver cDNA library in lambda ZAP revealed several clones for the mRNA of glutamine synthase. The longest clone was completely sequenced and consists of a 109 bp 5' untranslated region, a 1119 bp protein coding region, a 1498 bp 3' untranslated region and a poly(A) tract of 12 bp.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
L-Glutamine Synthetase from Escherichia coli, lyophilized powder, 400-2,000 units/mg protein