Merck
  • Homozygous tyrosinase gene mutation in an American black with tyrosinase-negative (type IA) oculocutaneous albinism.
Homozygous tyrosinase gene mutation in an American black with tyrosinase-negative (type IA) oculocutaneous albinism.
American journal of human genetics (1991-02-01)
R A Spritz, K M Strunk, C L Hsieh, G S Sekhon, U Francke
ZUSAMMENFASSUNG
We have identified a tyrosinase gene mutation in an American black with classic, tyrosinase-negative oculocutaneous albinism. This mutation results in an amino acid substitution (Cys----Arg) at codon 89 of the tyrosinase polypeptide. The proband is homozygous for the substitution, suggesting that this mutation may be frequently associated with tyrosinase-negative oculocutaneous albinism in blacks.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Tyrosinase aus Ständerpilz, lyophilized powder, ≥1000 unit/mg solid