Merck
Autoimmunity in dry eye is due to resistance of Th17 to Treg suppression.
Journal of immunology (Baltimore, Md. : 1950) (2009-01-22)
Sunil K Chauhan, Jaafar El Annan, Tatiana Ecoiffier, Sunali Goyal, Qiang Zhang, Daniel R Saban, Reza Dana
ZUSAMMENFASSUNG
Dry eye disease (DED), an inflammatory autoimmune disorder affecting the ocular surface, degrades visual performance and the quality of life of >10 million people in the United States alone. The primary limitation in the effective treatment of DED is an incomplete understanding of its specific cellular and molecular pathogenic elements. Using a validated mouse model of DED, herein we functionally characterize the different T cell subsets, including regulatory T cells (Tregs) and pathogenic effector T cells, and determine their contribution to the pathogenesis of DED. Our data demonstrate the presence of dysfunctional Tregs and the resistance of pathogenic T cells, particularly Th17 cells, to Treg suppression in DED. In addition, we clearly show that in vivo blockade of IL-17 significantly reduces the severity and progression of disease, which is paralleled by a reduction in the expansion of Th17 cells and restoration of Treg function. Our findings elucidate involvement of a previously unknown pathogenic T cell subset (Th17) in DED that is associated specifically with Treg dysfunction and disease pathogenesis and suggest a new target for dry eye therapy.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ionomycin aus Streptomyces conglobatus, ≥98% (HPLC)
Sigma-Aldrich
Scopolamin -hydrobromid, meets USP testing specifications