Merck
  • IL-1 mediates pulmonary and systemic inflammatory responses to chorioamnionitis induced by lipopolysaccharide.
IL-1 mediates pulmonary and systemic inflammatory responses to chorioamnionitis induced by lipopolysaccharide.
American journal of respiratory and critical care medicine (2009-02-24)
Suhas G Kallapur, Ilias Nitsos, Timothy J M Moss, Graeme R Polglase, J Jane Pillow, Fook-Choe Cheah, Boris W Kramer, John P Newnham, Machiko Ikegami, Alan H Jobe
ZUSAMMENFASSUNG
Chorioamnionitis frequently associates with preterm delivery and increased amniotic fluid IL-1, and causes fetal lung and systemic inflammation. However, chorioamnionitis is also associated with a paradoxical reduction in the incidence of surfactant deficiency-related respiratory distress syndrome in preterm infants. To identify the role of IL-1 signaling in the mediation of pulmonary and systemic inflammation and lung maturation in a fetal sheep model of lipopolysaccharide (LPS) induced chorioamnionitis. After confirming the efficacy of recombinant human IL-1 receptor antagonist (rhIL-1ra), fetal sheep were exposed to intraamniotic (IA) injections of Escherichia coli LPS with or without prior IA injections of rhIL-1ra. Preterm lambs were delivered at 82% of term gestation. rhIL-1ra decreased IA LPS-induced lung inflammation assessed by decreased lung neutrophil and monocyte influx, inducible nitric oxide synthase expression, lung IL-6 and IL-1beta mRNA expression, and airway myeloperoxidase concentrations. rhIL-1ra inhibited IA LPS-induced fetal systemic inflammation assessed by decreased plasma IL-8, protein carbonyls, blood neutrophilia, and the expression of serum amyloid A3 mRNA in the liver. rhIL-1ra also partially blocked the lung maturational effects of IA LPS. Therefore blockade of IL-1 signaling in the amniotic compartment inhibited fetal lung and systemic inflammation and lung maturation in response to LPS-induced chorioamnionitis. IL-1 plays a central role in the pathogenesis of chorioamnionitis-induced fetal inflammatory responses.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Lipopolysaccharide aus Escherichia coli O55:B5, purified by phenol extraction
Sigma-Aldrich
Lipopolysaccharide aus Escherichia coli O55:B5, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Lipopolysaccharide aus Escherichia coli O55:B5, purified by gel-filtration chromatography
Sigma-Aldrich
Lipopolysaccharide aus Escherichia coli O55:B5, Ready Made solution, 1 mg/mL
Sigma-Aldrich
Lipopolysaccharide aus Escherichia coli O55:B5, purified by ion-exchange chromatography, TLR ligand tested
Sigma-Aldrich
Lipopolysaccharide aus Escherichia coli O55:B5, purified by trichloroacetic acid extraction
Sigma-Aldrich
Lipopolysaccharide aus Escherichia coli O55:B5, FITC conjugate