Merck
The previously reported T342P GCK missense variant is not a pathogenic mutation causing MODY.
Diabetologia (2011-05-24)
A M Steele, N D Tribble, R Caswell, K J Wensley, A T Hattersley, A L Gloyn, S Ellard