Merck
  • Hepatoprotective Effects of Orthosiphon stamineus Extract on Thioacetamide-Induced Liver Cirrhosis in Rats.
Hepatoprotective Effects of Orthosiphon stamineus Extract on Thioacetamide-Induced Liver Cirrhosis in Rats.
Evidence-based complementary and alternative medicine : eCAM (2011-06-08)
Mohammed A Alshawsh, Mahmood Ameen Abdulla, Salmah Ismail, Zahra A Amin
ZUSAMMENFASSUNG
Orthosiphon stamineus as medicinal plant is commonly used in Malaysia for treatment of hepatitis and jaundice; in this study, the ethanol extracts were applied to evaluate the hepatoprotective effects in a thioacetamide-induced hepatotoxic model in Sprague Dawley rats. Five groups of adult rats were arranged as follows: Group 1 (normal control group), Group 2 Thioacetamide (TAA) as positive control (hepatotoxic group), Group 3 Silymarin as a well-known standard drug (hepatoprotective group), and Groups 4 and 5 as high and low dose (treatment groups). After 60-day treatment, all rats were sacrificed. The hepatotoxic group showed a coarse granulation on the liver surface when compared to the smooth aspect observed on the liver surface of the other groups. Histopathological study confirmed the result; moreover, there was a significant increase in serum liver biochemical parameters (ALT, AST, ALP, and Bilirubin) and the level of liver Malondialdehyde (MDA), accompanied by a significant decrease in the level of total protein and Albumin in the TAA control group when compared with that of the normal group. The high-dose treatment group (200 mg/kg) significantly restored the elevated liver function enzymes near to normal. This study revealed that 200 mg/kg extracts of O. stamineus exerted a hepatoprotective effect.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Thioacetamid, ACS reagent, ≥99.0%
Sigma-Aldrich
Thioacetamid, reagent grade, 98%