Merck
  • Genome-wide dFOXO targets and topology of the transcriptomic response to stress and insulin signalling.
Genome-wide dFOXO targets and topology of the transcriptomic response to stress and insulin signalling.
Molecular systems biology (2011-06-23)
Nazif Alic, T Daniel Andrews, Maria E Giannakou, Irene Papatheodorou, Cathy Slack, Matthew P Hoddinott, Helena M Cochemé, Eugene F Schuster, Janet M Thornton, Linda Partridge
ZUSAMMENFASSUNG
FoxO transcription factors, inhibited by insulin/insulin-like growth factor signalling (IIS), are crucial players in numerous organismal processes including lifespan. Using genomic tools, we uncover over 700 direct dFOXO targets in adult female Drosophila. dFOXO is directly required for transcription of several IIS components and interacting pathways, such as TOR, in the wild-type fly. The genomic locations occupied by dFOXO in adults are different from those observed in larvae or cultured cells. These locations remain unchanged upon activation by stresses or reduced IIS, but the binding is increased and additional targets activated upon genetic reduction in IIS. We identify the part of the IIS transcriptional response directly controlled by dFOXO and the indirect effects and show that parts of the transcriptional response to IIS reduction do not require dfoxo. Promoter analyses revealed GATA and other forkhead factors as candidate mediators of the indirect and dfoxo-independent effects. We demonstrate genome-wide evolutionary conservation of dFOXO targets between the fly and the worm Caenorhabditis elegans, enriched for a second tier of regulators including the dHR96/daf-12 nuclear hormone receptor.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
GenomePlex® Kit zur Gesamtgenom-Amplifikation (GGA), Kit for whole genome amplification from a variety of DNA sources including FFPE tissue