Merck
Two point mutations are responsible for G6PD polymorphism in Sardinia.
American journal of human genetics (1989-02-01)
G De Vita, M Alcalay, M Sampietro, M D Cappelini, G Fiorelli, D Toniolo
ZUSAMMENFASSUNG
The human X-linked gene encoding glucose 6-phosphate dehydrogenase (G6PD) is highly polymorphic; more than 300 G6PD variants have been identified. G6PD deficiency in different geographical areas appears to have arisen through independent mutational events, but within the same population it may also be heterogeneous. One example is the island of Sardinia, where careful clinical and biochemical studies have identified four different G6PD variants. We cloned and sequenced the four G6PD variants from Sardinia and found that only two mutations are responsible for G6PD deficiency in this area: one mutation is the cause of the G6PD Seattle-like phenotype, a milder form of G6PD deficiency; the other mutation is responsible for all forms of very severe G6PD deficiency in Sardinia and, possibly, in the Mediterranean.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Glucose-6-phosphat-Dehydrogenase aus Backhefe (S. cerevisiae), Type XV, lyophilized powder, 200-400 units/mg protein (modified Warburg-Christian)
Sigma-Aldrich
Glucose-6-phosphat-Dehydrogenase aus Backhefe (S. cerevisiae), Type VII, ammonium sulfate suspension, ≥200 units/mg protein
Sigma-Aldrich
Glucose-6-phosphat-Dehydrogenase aus Leuconostoc mesenteroides, recombinant, expressed in E. coli, ammonium sulfate suspension, ≥550 units/mg protein (biuret)
Sigma-Aldrich
Glucose-6-phosphat-Dehydrogenase aus Leuconostoc mesenteroides, lyophilized powder, >= 550 units/mg protein (biuret)
Sigma-Aldrich
Glucose-6-phosphat-Dehydrogenase aus Backhefe (S. cerevisiae), Type IX, lyophilized powder, 200-400 units/mg protein (modified Warburg-Christian)
Sigma-Aldrich
Glucose-6-phosphat-Dehydrogenase aus Leuconostoc mesenteroides, recombinant, expressed in E. coli, lyophilized powder, ≥550 units/mg protein (biuret)
Sigma-Aldrich
Glucose-6-phosphat-Dehydrogenase aus Leuconostoc mesenteroides, Type XXIII, ammonium sulfate suspension, 550-1,100 units/mg protein (biuret), ≥2.0 mg/mL Biuret