Merck
  • Purification and properties of N-acyl-D-mannosamine dehydrogenase from Flavobacterium sp. 141-8.
Purification and properties of N-acyl-D-mannosamine dehydrogenase from Flavobacterium sp. 141-8.
Journal of biochemistry (1988-09-01)
T Horiuchi, T Kurokawa
ZUSAMMENFASSUNG
A new enzyme, N-acyl-D-mannosamine dehydrogenase, was purified to apparent homogeneity from a cell-free extract of Flavobacterium sp. 141-8 and some of its properties were investigated. The enzyme showed optimum activity at pH 8.0-9.5. N-Acetyl- and N-glycolyl-D-mannosamine were oxidized but other commonly existing sugars, such as N-acetylglucosamine, N-acetylgalactosamine, amino sugars, neutral hexoses, and pentoses, were not oxidized. NAD+ was specifically utilized as an effective hydrogen acceptor. The apparent Km values for N-acetyl- and N-glycolyl-D-mannosamine, and NAD+ were 1.0, 13.3, and 0.41 mM, respectively. The stoichiometry data showed that 1 mol each of N-acetyl-D-mannosamine and NAD+ were converted to 1 mol each of N-acetyl-D-mannosaminic acid and NADH, respectively. Although the formation of lactone was detected in the enzyme reaction mixture, the reverse reaction of the enzyme, the reduction of N-acetyl-D-mannosamino-lactone, was not observed. The enzyme activity was strongly inhibited by Hg2+ and SDS, but metal-chelating reagents and sulfhydryl-group-blocking reagents had almost no effect. The molecular weight of the enzyme was estimated to be 120,000 on gel filtration and 29,000 on SDS-polyacrylamide gel electrophoresis. Its isoelectric point was at pH 4.8. On trial application of the enzyme, it was indicated that N-acetylneuraminic acid can be determined quantitatively with the combined enzyme system involving the new enzyme and N-acetylneuraminic acid aldolase.