- Enhanced expression of a glyceraldehyde-3-phosphate dehydrogenase gene in human lung cancers.
Enhanced expression of a glyceraldehyde-3-phosphate dehydrogenase gene in human lung cancers.
Cancer research (1987-11-01)
K Tokunaga, Y Nakamura, K Sakata, K Fujimori, M Ohkubo, K Sawada, S Sakiyama
PMID3664468
ZUSAMMENFASSUNG
We have shown that a Mr 37,000 protein whose expression is strongly enhanced in human lung cancer tissues is the subunit of glyceraldehyde-3-phosphate (GAPDH, EC 1.2.1.12). We have isolated a GAPDH complementary DNA from human lung cancer cells and deduced the complete amino acid sequence of the encoded protein. The structure of the lung cancer GAPDH is identical to that of liver GAPDH. In addition, we have found that GAPDH mRNA expression is markedly increased in human lung cancer tissues. These results disclose a molecular basis of increased glycolysis in cancer cells and reveal an important role of energy creating reaction in cancer cell growth.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Glyceraldehyd-3-Phosphat-Dehydrogenase aus Kaninchenmuskel, lyophilized powder, ≥75 units/mg protein
Sigma-Aldrich
Glyceraldehyde-3-phosphate Dehydrogenase from human erythrocytes, lyophilized powder, 50-150 units/mg protein