Merck
  • Glucose 6-phosphate dehydrogenase from human erythrocytes: identification of N-acetyl-alanine at the N-terminus of the mature protein.
Glucose 6-phosphate dehydrogenase from human erythrocytes: identification of N-acetyl-alanine at the N-terminus of the mature protein.
Biochemical and biophysical research communications (1995-02-06)
L Camardella, G Damonte, V Carratore, U Benatti, M Tonetti, G Moneti
ZUSAMMENFASSUNG
Glucose 6-phosphate dehydrogenase from human erythrocytes has a blocked amino-terminus and no information could be obtained by direct sequencing of the intact protein. The peptide corresponding to the amino-terminal region was isolated from a tryptic digest of the whole protein and identified on the basis of its amino acid composition and of the failure to obtain Edman degradation. Determination of peptide mass by fast atom bombardment mass spectrometry allowed identification of the blocked amino-terminal residue as N-acetyl-alanine.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Glucose-6-phosphat-Dehydrogenase aus Backhefe (S. cerevisiae), Type XV, lyophilized powder, 200-400 units/mg protein (modified Warburg-Christian)
Sigma-Aldrich
Glucose-6-phosphat-Dehydrogenase aus Backhefe (S. cerevisiae), Type VII, ammonium sulfate suspension, ≥200 units/mg protein
Sigma-Aldrich
Glucose-6-phosphat-Dehydrogenase aus Leuconostoc mesenteroides, recombinant, expressed in E. coli, ammonium sulfate suspension, ≥550 units/mg protein (biuret)
Sigma-Aldrich
Glucose-6-phosphat-Dehydrogenase aus Leuconostoc mesenteroides, lyophilized powder, >= 550 units/mg protein (biuret)
Sigma-Aldrich
Glucose-6-phosphat-Dehydrogenase aus Backhefe (S. cerevisiae), Type IX, lyophilized powder, 200-400 units/mg protein (modified Warburg-Christian)
Sigma-Aldrich
N-Acetyl-L-alanine, ~99%
Sigma-Aldrich
Glucose-6-phosphat-Dehydrogenase aus Leuconostoc mesenteroides, recombinant, expressed in E. coli, lyophilized powder, ≥550 units/mg protein (biuret)
Sigma-Aldrich
Glucose-6-phosphat-Dehydrogenase aus Leuconostoc mesenteroides, Type XXIII, ammonium sulfate suspension, 550-1,100 units/mg protein (biuret), ≥2.0 mg/mL Biuret