Merck
  • A microdialysis study of the effects of the nicotinic agonist RJR-2403 on cortical release of acetylcholine and biogenic amines.
A microdialysis study of the effects of the nicotinic agonist RJR-2403 on cortical release of acetylcholine and biogenic amines.
Neurochemical research (1996-10-01)
K L Summers, P Lippiello, E Giacobini
ZUSAMMENFASSUNG
Transcortical dialysis was employed to investigate the effects of subcutaneous (s.c.) injections of RJR-2403 (1.2-7.2 mumol/kg) on extracellular levels of acetylcholine (ACh), norepinephrine (NE), dopamine (DA), and serotonin (5-HT) in rat. Systemic administration of RJR-2403 produced a 90% increase of cortical extracellular ACh levels that persisted for up to 90 minutes after injection. Norepinephrine and DA release were increased 124% and 131% above basal values, respectively. Serotonin (5-HT) levels in the dialysate were also significantly elevated by RJR-2403 (3.6 mumol/kg, s.c.) 70% above baseline at 90 minutes post-injection. Comparison of these responses to those of (-)nicotine from a previous study reveals little difference between the two compounds in their ability to influence cortical neurotransmitter release following systemic administration.