Merck
PKC-dependent activation of human K(2P) 18.1 K(+) channels.
British journal of pharmacology (2011-12-16)
Ann-Kathrin Rahm, Jakob Gierten, Jana Kisselbach, Ingo Staudacher, Kathrin Staudacher, Patrick A Schweizer, Rüdiger Becker, Hugo A Katus, Dierk Thomas
ZUSAMMENFASSUNG
Two-pore-domain K(+) channels (K(2P) ) mediate K(+) background currents that modulate the membrane potential of excitable cells. K(2P) 18.1 (TWIK-related spinal cord K(+) channel) provides hyperpolarizing background currents in neurons. Recently, a dominant-negative loss-of-function mutation in K(2P) 18.1 has been implicated in migraine, and activation of K(2P) 18.1 channels was proposed as a therapeutic strategy. Here we elucidated the molecular mechanisms underlying PKC-dependent activation of K(2P) 18.1 currents. Human K(2P) 18.1 channels were heterologously expressed in Xenopus laevis oocytes, and currents were recorded with the two-electrode voltage clamp technique. Stimulation of PKC using phorbol 12-myristate-13-acetate (PMA) activated the hK(2P) 18.1 current by 3.1-fold in a concentration-dependent fashion. The inactive analogue 4α-PMA had no effect on channel activity. The specific PKC inhibitors bisindolylmaleimide I, Ro-32-0432 and chelerythrine reduced PMA-induced channel activation indicating that PKC is involved in this effect of PMA. Selective activation of conventional PKC isoforms with thymeleatoxin (100 nM) did not reproduce K(2P) 18.1 channel activation. Current activation by PMA was not affected by pretreatment with CsA (calcineurin inhibitor) or KT 5720 (PKA inhibitor), ruling out a significant contribution of calcineurin or cross-talk with PKA to the PKC-dependent hK(2P) 18.1 activation. Finally, mutation of putative PKC phosphorylation sites did not prevent PMA-induced K(2P) 18.1 channel activation. We demonstrated that activation of hK(2P) 18.1 (TRESK) by PMA is mediated by PKC stimulation. Hence, PKC-mediated activation of K(2P) 18.1 background currents may serve as a novel molecular target for migraine treatment.