Merck
  • Disruption of Abi1/Hssh3bp1 expression induces prostatic intraepithelial neoplasia in the conditional Abi1/Hssh3bp1 KO mice.
Disruption of Abi1/Hssh3bp1 expression induces prostatic intraepithelial neoplasia in the conditional Abi1/Hssh3bp1 KO mice.
Oncogenesis (2012-01-01)
X Xiong, A Chorzalska, P M Dubielecka, J R White, Y Vedvyas, C V Hedvat, A Haimovitz-Friedman, J A Koutcher, J Reimand, G D Bader, J A Sawicki, L Kotula
ZUSAMMENFASSUNG
Prostate cancer is one of the leading causes of cancer-related deaths in the United States and a leading diagnosed non-skin cancer in American men. Genetic mutations underlying prostate tumorigenesis include alterations of tumor suppressor genes. We tested the tumor suppressor hypothesis for ABI1/hSSH3BP1 by searching for gene mutations in primary prostate tumors from patients, and by analyzing the consequences of prostate-specific disruption of the mouse Abi1/Hssh3bp1 ortholog. We sequenced the ABI1/hSSH3BP1 gene and identified recurring mutations in 6 out of 35 prostate tumors. Moreover, complementation and anchorage-independent growth, proliferation, cellular adhesion and xenograft assays using the LNCaP cell line, which contains a loss-of-function Abi1 mutation, and a stably expressed wild-type or mutated ABI gene, were consistent with the tumor suppressor hypothesis. To test the hypothesis further, we disrupted the gene in the mouse prostate by breeding the Abi1 floxed strain with the probasin promoter-driven Cre recombinase strain. Histopathological evaluation of mice indicated development of prostatic intraepithelial neoplasia (PIN) in Abi1/Hssh3bp1 knockout mouse as early as the eighth month, but no progression beyond PIN was observed in mice as old as 12 months. Observed decreased levels of E-cadherin, β-catenin and WAVE2 in mouse prostate suggest abnormal cellular adhesion as the mechanism underlying PIN development owing to Abi1 disruption. Analysis of syngeneic cell lines point to the possibility that upregulation of phospho-Akt underlies the enhanced cellular proliferation phenotype of cells lacking Abi1. This study provides proof-of-concept for the hypothesis that Abi1 downregulation has a role in the development of prostate cancer.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
JumpStart Taq-DNA-Polymerase, with MgCl2