Merck
New chlorinated diphenyl ethers from an Aspergillus species.
Journal of natural products (2002-01-26)
Jaih Hargreaves, Ja-on Park, Emilio L Ghisalberti, Krishnapillai Sivasithamparam, Brian W Skelton, Allan H White
ZUSAMMENFASSUNG
Two new chlorinated diphenyl ethers (5, 6) have been isolated from the culture broth of an Aspergillus species obtained from leaf litter, together with the known benzophenone sulochrin (1), the grisandiene geodin (2), and the diphenyl ether asterric acid (3). The structure of another metabolite, methyl asterrate (4), was confirmed by single-crystal X-ray structure analysis.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Asterric acid, ≥95% (LC/MS-ELSD)