Merck
  • Binding of substituted and conformationally restricted derivatives of N-(3-phenyl-n-propyl)-1-phenyl-2-aminopropane at sigma-receptors.
Binding of substituted and conformationally restricted derivatives of N-(3-phenyl-n-propyl)-1-phenyl-2-aminopropane at sigma-receptors.
Journal of medicinal chemistry (1991-06-01)
R A Glennon, A M Ismaiel, J D Smith, M Yousif, M el-Ashmawy, J L Herndon, J B Fischer, K J Howie, A C Server
ZUSAMMENFASSUNG
Certain benzomorphans, such as N-allylnormetazocine, are classical "sigma-opiates" that bind both at sigma and phencyclidine (PCP) binding sites with modest affinity. Recently, we identified N-substituted 2-phenylaminoethane as being the primary sigma-pharmacophore of the benzomorphans and demonstrated that 1-phenyl-2-aminopropane (2) derivatives, depending upon their terminal amine substituents, constitute a novel class of high-affinity sigma-selective agents. With this pharmacophore, it is shown in the present investigation that the aromatic hydroxyl group (a prime feature of all the sigma-opiates) contributes little to the binding of 2 at sigma-sites. It is also demonstrated that an N-substituted aminotetralin moiety (such as 17, a conformationally restricted analogue of 2) may also be considered a sigma-opiate pharmacophore. Unlike the sigma-opiates, derivatives of 2 and 17 display no affinity for PCP sites and must consequently lack those structural features important for the binding of benzomorphans at PCP sites. Because 3-phenylpiperidines and related sigma-ligands also possess a phenylalkylamine imbedded within their structures, we propose that the 2-phenylaminoethane moiety is a common sigma-pharmacophore for derivatives of 2, the 3-phenylpiperidines, and the sigma-opiates.