Merck
  • Rhizoxin, a macrocyclic lactone antibiotic, as a new antitumor agent against human and murine tumor cells and their vincristine-resistant sublines.
Rhizoxin, a macrocyclic lactone antibiotic, as a new antitumor agent against human and murine tumor cells and their vincristine-resistant sublines.
Cancer research (1986-01-01)
T Tsuruo, T Oh-hara, H Iida, S Tsukagoshi, Z Sato, I Matsuda, S Iwasaki, S Okuda, F Shimizu, K Sasagawa
ZUSAMMENFASSUNG
Rhizoxin, isolated from a plant pathogenic fungus which causes rice seedling blight, inhibits the mitosis of the tumor cells in a manner similar to that of Vinca alkaloids as revealed by morphological study and flow cytometry analysis. This new 16-membered macrocyclic lactone showed similar chemotherapeutic effects to those of vincristine against L1210 and P388 leukemia-bearing mice. The drug is also effective against B16 melanoma inoculated i.p. or s.c. Rhizoxin, in contrast to the ansamacrolide, maytansine, was effective against human and murine tumor cells resistant to vincristine and Adriamycin in vitro and in vivo. A maximum 60% increase in life span was obtained in mice inoculated with P388 leukemia resistant to vincristine. Rhizoxin showed greater cytotoxicity in cultured tumor cells than did vincristine. Rhizoxin seems to bear consideration for further development as a new chemotherapeutic agent.