Merck
  • Antagonism of nicotinic receptors of rat chromaffin cells by N,N, N-trimethyl-1-(4-trans-stilbenoxy)-2-propylammonium iodide: a patch clamp and ligand binding study.
Antagonism of nicotinic receptors of rat chromaffin cells by N,N, N-trimethyl-1-(4-trans-stilbenoxy)-2-propylammonium iodide: a patch clamp and ligand binding study.
British journal of pharmacology (2000-04-26)
S Di Angelantonio, A Nistri, M Moretti, F Clementi, C Gotti
ZUSAMMENFASSUNG
The effect of the oxystilbene derivative F3 was tested on nAChRs of whole-cell patch-clamped rat chromaffin cells in vitro and of rat adrenal gland membranes using (125)I-epibatidine. F3 (30 nM) rapidly and reversibly blocked inward currents generated by pulse applications of nicotine, shifting the dose-response curve to the right in a parallel fashion without changing the maximum response. The action of F3 was voltage insensitive and not due to altered current reversal potential. The R isomer of F3 was more potent (IC(50) = 350+/-30 nM) than its S-enantiomer (IC(50) = 1.5+/-0.3 microM). Nicotine-evoked currents were insensitive to 10 microM alpha-bungarotoxin. Equi-amplitude currents evoked by nicotine or epibatidine were similarly antagonized by R-F3 in a reversible fashion. Epibatidine-evoked currents readily produced receptor desensitization. Adrenal membranes specifically bound (125)I-epibatidine with a single population of binding sites endowed with high affinity (K(D) = 159 pM) and B(max) of 6.5+/-1.3 fmol mg(-1) of protein. (125)I-epibatidine binding was specifically displaced by cytisine (K(i) = 68 nM) or ACh (K(i) = 348 nM). F3 specifically displaced (125)I-epibatidine binding although with lower affinity (K(i) = 29.6 microM) than in electrophysiological experiments. (125)I-epibatidine binding to rat adrenal tissue was insensitive to alpha-bungarotoxin which readily antagonized (125)I-epibatidine binding to bovine adrenal tissue. The present results suggest that F3 is a relatively potent and apparently competitive antagonist of nAChRs on rat chromaffin cells. Since previous studies have indicated that F3 targets different subtypes on chick neuronal tissue, it appears that nAChRs display interspecies differences to be considered for drug development studies.