Merck
  • JNK1: a protein kinase stimulated by UV light and Ha-Ras that binds and phosphorylates the c-Jun activation domain.
JNK1: a protein kinase stimulated by UV light and Ha-Ras that binds and phosphorylates the c-Jun activation domain.
Cell (1994-03-25)
B Dérijard, M Hibi, I H Wu, T Barrett, B Su, T Deng, M Karin, R J Davis
ZUSAMMENFASSUNG
The ultraviolet (UV) response of mammalian cells is characterized by a rapid and selective increase in gene expression mediated by AP-1 and NF-kappa B. The effect on AP-1 transcriptional activity results, in part, from enhanced phosphorylation of the c-Jun NH2-terminal activation domain. Here, we describe the molecular cloning and characterization of JNK1, a distant relative of the MAP kinase group that is activated by dual phosphorylation at Thr and Tyr during the UV response. Significantly, Ha-Ras partially activates JNK1 and potentiates the activation caused by UV. JNK1 binds to the c-Jun transactivation domain and phosphorylates it on Ser-63 and Ser-73. Thus, JNK1 is a component of a novel signal transduction pathway that is activated by oncoproteins and UV irradiation. These properties indicate that JNK1 activation may play an important role in tumor promotion.