Merck
Depletion rate of N4-acetylsulphamonomethoxine in tissues of laying hens.
Veterinary journal (London, England : 1997) (1999-02-25)
N Furusawa
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Sulfamonomethoxin, VETRANAL®, analytical standard
Sigma-Aldrich
Sulfamonomethoxin