Merck
  • Persistence of gentian violet and leucogentian violet in channel catfish (ictalurus punctatus) muscle after water-borne exposure.
Persistence of gentian violet and leucogentian violet in channel catfish (ictalurus punctatus) muscle after water-borne exposure.
Journal of chromatography. B, Biomedical sciences and applications (1999-03-18)
H C Thompson, L G Rushing, T Gehring, R Lochmann
ZUSAMMENFASSUNG
Gentian violet is a triphenylmethane dye that is an antifungal/antiparastic agent. GV is similar to malachite green that has been used in the aquaculture industry for treatment or prevention of external fungal and parasitic infections in fish and fish eggs although it (MG) is not approved for this use. For these reasons, GV's potential for misuse by the aquaculture industry is high. The uptake and depletion of gentian violet (GV) were determined in channel catfish (Ictalurus punctatus) after water-borne exposure (100 ng ml(-1), 1 h) under simulated aquaculture farming conditions. Leucogentian violet (LGV) was rapidly formed, concentrated in the muscle tissue, and very slowly eliminated from muscle tissue. An isocratic (60% acetonitrile-40% water; 0.05 M ammonium acetate buffer, pH 4.5) HPLC system consisting of a 5 microm LC-CN 250x4.6 mm I.D. column, a 20x2.0 mm I.D. PbO2 oxidative post-column, and a UV-VIS detector set at 588 nm were used to determine uptake and depletion of tissue residues of GV and LGV with time. GV was rapidly depleted and converted to its major metabolite, LGV, which was detected out to 79 days. Therefore, LGV is the appropriate target analyte for monitoring exposure of channel catfish to GV.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Leukokristallviolett