- [The possibility of using risk assessments of the development of diagnostically different body states for setting hygienic standards].
[The possibility of using risk assessments of the development of diagnostically different body states for setting hygienic standards].
Gigiena i sanitariia (1999-04-13)
T E Shimonov, S L Avaliani, M M Andrianova
PMID10199093
MATERIALIEN