Merck
  • Cerium-based histochemical demonstration of oxidative stress in taurocholate-induced acute pancreatitis in rats. A confocal laser scanning microscopic study.
Cerium-based histochemical demonstration of oxidative stress in taurocholate-induced acute pancreatitis in rats. A confocal laser scanning microscopic study.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society (1999-08-17)
G Telek, J Y Scoazec, J Chariot, R Ducroc, G Feldmann, C Roz
ZUSAMMENFASSUNG
Direct in vivo histological detection of oxygen-derived free radicals (OFRs) in inflammatory conditions is not fully resolved. We report an application of cerium histochemistry (in which capture of OFRs by Ce atoms results in laser-reflectant cerium-perhydroxide precipitates) combined with reflectance confocal laser scanning microscopy (CLSM) to demonstrate the evolution of oxidative stress in taurocholate-induced acute pancreatitis (AP) in rats. Animals were perfused with CeCl(3) in vivo and cryostat sections of pancreata were studied by CLSM. Vascular endothelium was immunolabeled for PECAM-1. OFR production by isolated polymorphonuclear leukocytes (PMNs) incubated in vitro with CeCl(3) was quantified by image analysis. In the pancreas, strong OFR-derived cerium reflectance signals were seen in acinar cells at 1-2 hr, capillaries and small venules were frequently engorged by cerium precipitates, and adherent PMNs presented weak intracellular reflectance signals. At 8-24 hr, acinar cell OFR production decreased, whereas adherent/transmigrated PMNs displayed abundant intra- and pericellular reflectance. PECAM-1 expression was unchanged. PMNs from ascites or blood showed significant (p<0.01) time-dependent OFR production, plateauing from 2 hr. The modified cerium capture/CLSM method allows the co-demonstration of in vivo oxidative stress and cellular structures labeled with fluorescent markers. In vivo oxidative stress was shown histologically for the first time in experimental AP.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Cer(IV)-hydroxid