- Outer membrane proteins Omp10, Omp16, and Omp19 of Brucella spp. are lipoproteins.
Outer membrane proteins Omp10, Omp16, and Omp19 of Brucella spp. are lipoproteins.
Infection and immunity (1999-08-24)
A Tibor, B Decelle, J J Letesson
PMID10456959
ZUSAMMENFASSUNG
The deduced sequences of the Omp10, Omp16, and Omp19 outer membrane proteins of Brucella spp. contain a potential bacterial lipoprotein processing sequence. After extraction with Triton X-114, these three proteins partitioned into the detergent phase. Processing of the three proteins is inhibited by globomycin, a specific inhibitor of lipoprotein signal peptidase. The three proteins were radioimmunoprecipitated from [(3)H]palmitic acid-labeled Brucella abortus lysates with monoclonal antibodies. These results demonstrate that Omp10, Omp16, and Omp19 are lipoproteins.
MATERIALIEN