Merck
Water determination in drugs containing thiols.
International journal of pharmaceutics (1999-11-05)
F Sherman, I Kuselman
ZUSAMMENFASSUNG
A new rapid analytical method is applied for water determination in alpha-Mono-thioglycerol and Captopril tablets containing thiols, and therefore, not amenable for direct Karl Fischer titration. The method is based on the consecutive titration first of thiol by a novel reagent, and then of water by a conventional K. Fischer reagent in the same sample and cell with the electrometric 'dead-stop' location of the end point in both titrations. The new reagent consists of iodine, potassium iodide and sodium acetate in non-aqueous medium. Estimated repeatability and accuracy of both water and thiol determinations are satisfactory.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
1-Thioglycerin, liquid, BioReagent, suitable for cell culture, ≥97% (titration)
Sigma-Aldrich
1-Thioglycerin, ≥99.0% (GC)
Sigma-Aldrich
1-Thioglycerin, ≥97%
Sigma-Aldrich
1-Thioglycerin, BioXtra, ≥97%