- Site-specific formation of abasic lesions in DNA.
Site-specific formation of abasic lesions in DNA.
Nucleosides & nucleotides (2000-01-05)
R S Coleman, R M Pires
PMID10616722
ZUSAMMENFASSUNG
A method for the introduction of depurinated lesions in DNA is described, and is based on the formation of a covalent cross-link between an antisense oligonucleotide probe and the target DNA sequence followed by an unexpectedly mild thermal depurination.
MATERIALIEN