Merck
  • Simultaneous plasma level analysis of clomipramine, N-desmethylclomipramine, and fluvoxamine by reversed-phase liquid chromatography.
Simultaneous plasma level analysis of clomipramine, N-desmethylclomipramine, and fluvoxamine by reversed-phase liquid chromatography.
Therapeutic drug monitoring (2000-04-25)
L Palego, D Marazziti, L Biondi, G Giannaccini, N Sarno, A Armani, A Lucacchini, G B Cassano, L Dell'Osso
ZUSAMMENFASSUNG
A simple reversed-phase liquid chromatographic method enabling the simultaneous analysis in plasma of the tricyclic antidepressant clomipramine, its demethylated metabolite, and the selective serotonin reuptake inhibitor fluvoxamine, was developed. The drugs and dibenzepine, the internal standard, were extracted from 1 mL plasma through an automated solid-phase procedure, eluted in a total chromatographic time of approximately 14 min and detected by means of an ultraviolet spectrophotometer preset at 254 nm. An assay sensitivity of 10 microg/L was observed for all analytes. Recoveries for these drugs and their metabolites ranged between 65% and 98% and their coefficient of variation (within-day and day-to-day) between 1.9% and 2.9%. In spiked plasma, within-day and day-to-day imprecision data were less than 5%. The simultaneous determination of clomipramine, N-desmethylclomipramine, and fluvoxamine with adequate sensitivity and accuracy may be useful for the monitoring of drug treatment in depression and obsessive-compulsive disorder, where combinations of such drugs are employed.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
N-Desmethylclomipramine hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Norclomipramin -hydrochlorid, ≥98% (TLC), powder