Merck
  • Interaction of 4-hydroxybiphenyl with cyclodextrins: effect of complex structure on spectroscopic and photophysical properties.
Interaction of 4-hydroxybiphenyl with cyclodextrins: effect of complex structure on spectroscopic and photophysical properties.
Chemistry (Weinheim an der Bergstrasse, Germany) (2000-06-06)
P Bortolus, G Marconi, S Monti, B Mayer, G Köhler, G Grabner
ZUSAMMENFASSUNG
The structures, spectroscopic and photophysical properties of the inclusion complexes between 4-OH-biphenyl and cyclodextrins (CD) in water were studied theoretically and experimentally. The complex structures were predicted using a dynamic Monte Carlo procedure including solvation effects and analyzed via computed and experimentally determined circular dichroism. The interpretation of the induced circular dichroism spectra indicated the formation of stable 2:2 complexes between the probe and alpha-CD, while 1:1 structures account for the circular dichroism induced by beta- and gamma-CD. Fluorescence emission quantum yields and lifetimes, measured as a function of the CD concentration, confirmed the formation of this higher order complex with alpha-CD. Prototropic equilibration in the singlet excited state was found to be depressed in 2:2 complexes due to the hydrophobic environment of the OH groups, while it remained unperturbed in 1:1 complexes, where the substituent is exposed to the aqueous environment. Triplet-triplet absorption and triplet quenching data supported this interpretation. The photophysical properties of both the 1:1 and the 2:2 complexes are characterized by a significant reduction of the nonradiative decay rates.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
4-Phenylphenol, 97%
Sigma-Aldrich
4-Phenylphenol, purified by sublimation, 99%