Merck
  • Structure/function of the human Ga1beta1,3-glucuronosyltransferase. Dimerization and functional activity are mediated by two crucial cysteine residues.
Structure/function of the human Ga1beta1,3-glucuronosyltransferase. Dimerization and functional activity are mediated by two crucial cysteine residues.
The Journal of biological chemistry (2000-06-08)
M Ouzzine, S Gulberti, P Netter, J Magdalou, S Fournel-Gigleux
ZUSAMMENFASSUNG
Galbeta1,3-glucuronosyltransferase (GlcAT-I) that catalyzes the transfer of a glucuronic acid residue onto the trisaccharide primer of the glycosaminoglycan-protein linkage region plays an essential role in the early steps of the biosynthesis of glycosaminoglycans. In order to gain insight into the structure/function of the enzyme, the human recombinant GlcAT-I was successfully expressed in the yeast Pichia pastoris, with an apparent molecular mass of 43 kDa. Analysis of the electrophoretic mobility of the membrane-bound protein in nonreducing and reducing conditions, together with cross-linking studies, indicated that the membrane-bound GlcAT-I formed active disulfide-linked dimers. GlcAT-I expressed without the predicted N-terminal cytoplasmic tail or secreted as a polypeptide lacking the cytoplasmic tail and transmembrane domain was similarly organized as dimers, suggesting that the structural determinants for the dimerization state are localized in the luminal domain of the protein. In addition, the role of Cys(33) and Cys(301) in that process was investigated by site-directed mutagenesis combined with chemical modification of GlcAT-I by N-phenylmaleimide. Replacement of Cys(33) with alanine abolished the formation of dimers with a concomitant decrease in the catalytic efficiency mainly due to a decrease in apparent maximal velocity and in affinity for UDP-glucuronic acid. On the other hand, N-phenylmaleimide treatment or alanine substitution of the Cys(301) residue inactivated the enzyme. Our study demonstrates that GlcAT-I is organized as a homodimer as a result of disulfide bond formation mediated by Cys(33) localized in the stem region, whereas the residue Cys(301) localized in a conserved C-terminal domain is strictly required for the functional integrity of the enzyme.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
N-Phenylmaleimid, 97%