Merck
  • Applications of plastic embedding to electron microscopic immunocytochemistry and in situ hybridization in observations of production and secretion of peptide hormones.
Applications of plastic embedding to electron microscopic immunocytochemistry and in situ hybridization in observations of production and secretion of peptide hormones.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society (2000-06-17)
R Y Osamura, Y Itoh, A Matsuno
ZUSAMMENFASSUNG
Plastic embedding has been used to localize various antigens in conjunction with immunohistochemistry. Peptide hormones have been among the antigens that have been studied extensively. Recent application of water-soluble plastics such as LR White and Lowicryl has extended the ranges of detectable antigens and enabled the observation of antigen-antigen or mRNA-antigen combinations. This review article deals with technical aspects, procedures, and applications in endocrine cells.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
LR weiße Acrylharze, Medium