Merck
A survey of Fusarium toxins in cereal-based foods marketed in an area of southwest Germany.
Mycopathologia (2000-06-29)
M Schollenberger, S Suchy, H T Jara, W Drochner, H M Müller
ZUSAMMENFASSUNG
A total of 237 commercially available samples of cereal-based foods including bread and related products, noodles, breakfast cereals, baby and infant foods, rice and other foods were randomly collected in southwest Germany during the first six months of 1998. The trichothecenes deoxynivalenol (DON), 3- and 15-acetyl-deoxynivalenol (3-,15-ADON), nivalenol (NIV), fusarenon-X (FUS-X), T-2 toxin (T-2) and HT-2 toxin (HT-2) were determined by gas chromatography/mass spectrometry following clean-up by a two stage solid-phase extraction. Detection limits ranged between 2 and 12 micrograms/kg. Based on all samples, the incidence of DON, HT-2, T-2, 3-ADON, 15-ADON, and NIV was at 71, 18, 4, 4, 4 and 2%, respectively; the average contents in positive samples were at 103, 16, 14, 17, 24 and 109 micrograms/kg, respectively. Fus-X was not detected in any sample. A lower (P < 0.05) DON content was found in baby and infant foods as well as in cookies and cakes compared to bread. Overall, based on the incidence and level of all six toxins, the degree of contamination was lowest in baby and infant foods. Foods produced from either white or whole grain flour did not differ (P > 0.05) with regard to the incidence and level of DON. In foods produced from cereals of organic production both the incidence and median content of DON was lower compared to conventional production. Zearalenone, alpha- and beta-zearalenol were determined by high performance liquid chromatography in 20 selected samples, mostly baby and infant foods. These toxins were not present in excess of the detection limit in any sample.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Fusarenon X -Lösung, 100 μg/mL in acetonitrile, analytical standard
Supelco
Fusarenon X, analytical standard