Merck
  • Phenytoin and its metabolite, 5-(4-hydroxyphenyl)-5-phenylhydantoin, show bone resorption in cultured neonatal mouse calvaria.
Phenytoin and its metabolite, 5-(4-hydroxyphenyl)-5-phenylhydantoin, show bone resorption in cultured neonatal mouse calvaria.
Japanese journal of pharmacology (2000-06-30)
A Takahashi, K Onodera, H Shinoda, H Mayanagi
ZUSAMMENFASSUNG
The effects of phenytoin and its major metabolite, 5-(4-hydroxyphenyl)-5-phenylhydantoin (HPPH), on bone resorption of neonatal mouse calvaria were examined in vitro. Both phenytoin and HPPH induced significant bone resorption as compared to the controls after 72 h in culture. This effect may be the cause of phenytoin-induced bone loss in vivo.