Merck
The location of the mobile electron carrier ferredoxin in vascular plant photosystem I.
The Journal of biological chemistry (2000-08-19)
S V Ruffle, A O Mustafa, A Kitmitto, A Holzenburg, R C Ford
ZUSAMMENFASSUNG
In this study, we present the location of the ferredoxin-binding site in photosystem I from spinach. Image analysis of negatively stained two-dimensional crystals indicates that the addition of ferredoxin and chemical cross-linkers do not significantly alter the unit cell parameters (for untreated photosystem I, a = 26.4 nm, b = 27.6 nm, and gamma = 90 degrees, space group p22(1)2(1) and for ferredoxin cross-linked photosystem I, a = 26.2 nm, b = 27.2 nm, and gamma = 90 degrees, space group p22(1)2(1)). Fourier difference analysis reveals that ferredoxin is bound on top of the stromal ridge principally interacting with the extrinsic subunits PsaC and PsaE. This location would be accessible to the stroma, thereby promoting efficient electron transfer away from photosystem I. This observation is significantly different from that of the ferredoxin binding site proposed for cyanobacteria. A model for the binding of ferredoxin in vascular plants is proposed and is discussed relative to observations in cyanobacteria.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
1-[3-(Dimethylamino)propyl]-3-ethylcarbodiimidmethiodid