Merck
  • The high-sensitivity determination of protein concentrations by the enhancement of Rayleigh light scattering of Arsenazo-DBN.
The high-sensitivity determination of protein concentrations by the enhancement of Rayleigh light scattering of Arsenazo-DBN.
The Analyst (2000-09-26)
Q Li, X Chen, H Zhang, C Xue, S Liu, Z Hu
ZUSAMMENFASSUNG
A new Rayleigh light scattering (RLS) assay of protein is presented in this paper. At the optimum pH 4.10, the weak RLS of Arsenazo-DBN can be greatly enhanced by the addition of proteins due to the interaction between protein and Arsenazo-DBN. Based on this, the reactions of Arsenazo-DBN and proteins, including bovine serum albumin, human serum album, gamma-globulin, egg albumin, lysozyme and trypsin, were studied. A new quantitative determination method for proteins has been developed. The linear range for human serum albumin, for example, is 0.085-34.62 micrograms mL-1 with a detection limit of 44.8 ng mL-1. Besides high sensitivity, the method is characterized by good reproducibility, rapidity of reaction, good stability, and few interfering substances. The determination of the proteins in human serum and urine samples by this method give results very close to those obtained using Coomassie Brilliant Blue G-250 colorimetry, with relative standard deviations of 0.7-2.5%.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Arsenazo III, calcium-sensitive dye