Merck
  • gamma-phosphate protonation and pH-dependent unfolding of the Ras.GTP.Mg2+ complex: a vibrational spectroscopy study.
gamma-phosphate protonation and pH-dependent unfolding of the Ras.GTP.Mg2+ complex: a vibrational spectroscopy study.
The Journal of biological chemistry (2001-01-02)
H Cheng, S Sukal, R Callender, T S Leyh
ZUSAMMENFASSUNG
The interdependence of GTP hydrolysis and the second messenger functions of virtually all GTPases has stimulated intensive study of the chemical mechanism of the hydrolysis. Despite numerous mutagenesis studies, the presumed general base, whose role is to activate hydrolysis by abstracting a proton from the nucleophilic water, has not been identified. Recent theoretical and experimental work suggest that the gamma-phosphate of GTP could be the general base. The current study investigates this possibility by studying the pH dependence of the vibrational spectrum of the Ras.GTP.Mg(2+) and Ras.GDP.Mg(2+) complexes. Isotope-edited IR studies of the Ras.GTP.Mg(2+) complex show that GTP remains bound to Ras at pH as low as 2.0 and that the gamma-phosphate is not protonated at pH > or = 3.3, indicating that the active site decreases the gamma-phosphate pK(a) by at least 1.1 pK(a) units compared with solution. Amide I studies show that the Ras.GTP.Mg(2+) and Ras.GDP.Mg(2+) complexes partially unfold in what appear to be two transitions. The first occurs in the pH range 5.4-2.6 and is readily reversible. Differences in the pH-unfolding midpoints for the Ras.GTP.Mg(2+) and Ras.GDP.Mg(2+) complexes (3.7 and 4.8, respectively) reveal that the enzyme-gamma-phosphoryl interactions stabilize the structure. The second transition, pH 2.6-1.7, is not readily reversed. The pH-dependent unfolding of the Ras.GTP.Mg(2+) complex provides an alternative interpretation of the data that had been used to support the gamma-phosphate mechanism, thereby raising the issue of whether this mechanism is operative in GTPase-catalyzed GTP hydrolysis reactions.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Guanosin 5′-triphosphat Natriumsalz Hydrat, ≥95% (HPLC), powder
Sigma-Aldrich
Guanosin-5′-triphosphat Natriumsalz Hydrat, ≥90% (HPLC)
Sigma-Aldrich
Guanosin-5′-triphosphat Natriumsalz -Lösung, HPLC purified, aqueous solution for RNA polymerase transcription