Merck
  • MAO-B inhibition by a single dose of l-deprenyl or lazabemide does not prevent neuronal damage following focal cerebral ischaemia in rats.
MAO-B inhibition by a single dose of l-deprenyl or lazabemide does not prevent neuronal damage following focal cerebral ischaemia in rats.
Pharmacology & toxicology (2000-12-29)
J Jolkkonen, R Kauppinen, L Nyman, A Haapalinna, J Sivenius
ZUSAMMENFASSUNG
The present study investigated the effect of postischaemic infusion of an irreversible monoamine oxidase B (MAO-B) inhibitor, l-deprenyl, an equipotent dose of a reversible MAO-B inhibitor, lazabemide, or 0.9% NaCl on infarct volumes following focal cerebral ischaemia in rats. The drug doses (0.3 mg/kg) were selected to induce selective MAO-B inhibition (45-55%), but not MAO-A inhibition. The infarct volumes in the cortex or in the striatum did not differ between the experimental groups 72 hr after transient occlusion of the middle cerebral artery, which suggests that during ischaemia/reperfusion, suppressed oxidative stress by partial MAO-B inhibition or MAO-B independent mechanisms such as induction of trophic factors, does not protect against ischaemia/reperfusion damage.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Lazabemide hydrate, ≥97% (HPLC)