Merck
  • Specific heterodimer formation by the cytoplasmic domains of the b and b' subunits of cyanobacterial ATP synthase.
Specific heterodimer formation by the cytoplasmic domains of the b and b' subunits of cyanobacterial ATP synthase.
Biochemistry (2001-01-05)
S D Dunn, E Kellner, H Lill
ZUSAMMENFASSUNG
The soluble domains of the b and b' subunits of the ATP synthase of the cyanobacterium Synechocystis PCC 6803 were expressed with His tags attached to their N-termini. Following purification, the polypeptides were characterized by chemical cross-linking, analytical ultracentrifugation, and circular dichroism spectroscopy. Treatment of a mixture of the soluble b and b' domains with a chemical cross-linking agent led to substantial formation of cross-linked dimers, whereas similar treatment of either domain by itself resulted in only trace formation of cross-linked species. The molecular weights of the domains of b and b' in solution at 20 degrees C, measured by sedimentation equilibrium, were 17 800+/-700 and 16 300+/-400, respectively, compared to calculated polypeptide molecular weights of 16 635 and 15 422, whereas a mixture of b and b' gave a molecular weight of 29 800+/-800. The sedimentation coefficient of an equimolar mixture was 1.73+/-0.03. The circular dichroism spectra of the individual polypeptides indicated helical contents in the range of 40-50%; the spectrum of the mixture revealed changes indicative of coiled-coil formation and a helical content of 60%. The results indicate that the cytosolic domains of the b and b' subunits exist individually as monomers but form a highly extended heterodimer when they are mixed together.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Suberic acid bis(3-sulfo-N-hydroxysuccinimide ester) sodium salt, ≥95% (H-NMR), powder