Merck
Chemiluminescence characteristics of N-(4-substituted benzyl)isoluminol.
Chemical & pharmaceutical bulletin (2001-01-06)
K Todoroki, Y Ohba, K Zaitsu
ZUSAMMENFASSUNG
We synthesized N-(4-substituted benzyl)isoluminol which has 4-bromo-, 4-methyl-, 4-methoxy-, 4-nitrogroups. These compounds produced chemiluminescence by the reaction with the oxidizing agent, potassium hexacyanoferrate and hydrogen peroxide, in an alkaline medium. The chemiluminescence intensities of these compounds were 0.03-4.7 times that of isoluminol. We used Hamett substituent constants as a parameter for the electronic substituent effects. The relationship between the amino-H chemical shift value and the Hamett substituent constants showed a good linear correlation. The relationship between the chemiluminescence intensities and the Hamett substituent constants showed a good linear correlation. The relationship between the fluorescence intensities and the Hamett substituent constants also showed a good linear correlation. These results suggest that the change in the electron density around the amino group strongly influences the fluorescence intensities and corresponding chemiluminescence intensities of these derivatives.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
4-Aminophthalhydrazid